Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077046.1 | NP_001070514.1 | 5 - 6 | 725 | 63 | |
GenBank mRNA | BC104531.1 | - | 5 - 6 | 725 | 63 | |
BT025413.1 | - | 5 - 6 | 703 | 63 | ||
BT026183.1 | - | 5 - 6 | 707 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |