Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HORMAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078143.2 | NP_001071611.1 | 4 - 5 | 313 | 121 | |
GenBank mRNA | BC120469.1 | - | 4 - 5 | 314 | 121 | |
BC150041.1 | - | 9 - 10 | 705 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |