Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174298.4 | NP_776723.1 | 2 - 3 | 315 | 111 | |
GenBank mRNA | AY033651.1 | - | 2 - 3 | 206 | 111 | |
BC102845.1 | - | 2 - 3 | 316 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |