Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024509.3 | NP_001019680.2 | 7 - 8 | 571 | 90 | |
GenBank mRNA | BC105379.1 | - | 7 - 8 | 572 | 90 | |
DQ010410.1 | - | 7 - 8 | 489 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |