Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037601.2 | NP_001032690.1 | 2 - 3 | 286 | 121 | |
GenBank mRNA | BC109556.1 | - | 2 - 3 | 287 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |