Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H11orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038037.2 | NP_001033126.1 | 5 - 6 | 506 | 106 | |
GenBank mRNA | BC109973.1 | - | 5 - 6 | 507 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |