Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038044.2 | NP_001033133.1 | 3 - 4 | 612 | 79 | |
GenBank mRNA | BC111283.1 | - | 3 - 4 | 613 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |