Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOT1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038058.1 | NP_001033147.1 | 1 - 2 | 229 | 124 | |
GenBank mRNA | BC111285.1 | - | 1 - 2 | 229 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |