Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038182.2 | NP_001033271.1 | 1 - 2 | 54 | 72 | |
GenBank mRNA | BC109620.1 | - | 1 - 2 | 55 | 72 | |
EF534775.1 | - | 1 - 2 | 45 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |