Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205485.1 | NP_001192414.1 | 6 - 7 | 636 | 109 | |
GenBank mRNA | BC111296.1 | - | 10 - 11 | 1149 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |