Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038685.2 | NP_001033774.1 | 6 - 7 | 586 | 82 | |
GenBank mRNA | BC110198.1 | - | 6 - 7 | 587 | 82 | |
BC114771.1 | - | 6 - 7 | 657 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |