Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040550.2 | NP_001035640.1 | 2 - 3 | 494 | 69 | |
GenBank mRNA | BC110021.1 | - | 2 - 3 | 495 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |