Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046005.2 | NP_001039470.1 | 1 - 2 | 326 | 110 | |
GenBank mRNA | BC112803.1 | - | 1 - 2 | 327 | 110 | |
FJ798978.1 | - | 1 - 2 | 189 | 110 | ||
FJ897536.1 | - | 1 - 2 | 189 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |