Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046048.1 | NP_001039513.1 | 8 - 9 | 990 | 103 | |
GenBank mRNA | BC105545.1 | - | 8 - 9 | 990 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |