Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046355.2 | NP_001039820.1 | 1 - 2 | 177 | 70 | |
GenBank mRNA | BC112884.1 | - | 1 - 2 | 178 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |