Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046479.2 | NP_001039944.1 | 6 - 7 | 535 | 76 | |
NM_001113242.2 | NP_001106713.1 | 8 - 9 | 1166 | 76 | ||
GenBank mRNA | BC110159.1 | - | 6 - 7 | 536 | 76 | |
BC147967.1 | - | 8 - 9 | 1168 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |