Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046502.2 | NP_001039967.1 | 4 - 5 | 1006 | 113 | |
GenBank mRNA | BC105377.1 | - | 4 - 5 | 1008 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |