Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075166.1 | NP_001068634.1 | 3 - 4 | 627 | 69 | |
GenBank mRNA | BC109588.1 | - | 3 - 4 | 627 | 69 | |
EU005652.1 | - | 3 - 4 | 650 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |