Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075763.2 | NP_001069231.1 | 7 - 8 | 1047 | 68 | |
GenBank mRNA | BC111238.1 | - | 7 - 8 | 1049 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |