Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB8OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076288.2 | NP_001069756.1 | 7 - 8 | 956 | 94 | |
GenBank mRNA | BC110257.1 | - | 7 - 8 | 957 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |