Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076500.2 | NP_001069968.1 | 3 - 4 | 331 | 69 | |
GenBank mRNA | BC109737.1 | - | 3 - 4 | 332 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |