Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076962.2 | NP_001070430.1 | 3 - 4 | 631 | 89 | |
GenBank mRNA | BC111227.1 | - | 3 - 4 | 632 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |