Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077084.2 | NP_001070552.1 | 1 - 2 | 109 | 138 | |
GenBank mRNA | BC111662.1 | - | 1 - 2 | 110 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |