Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077123.2 | NP_001070591.1 | 5 - 6 | 362 | 137 | |
GenBank mRNA | BC110123.1 | - | 5 - 6 | 363 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |