Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011676.3 | NP_001011676.2 | 4 - 5 | 751 | 100 | |
GenBank mRNA | AY841149.1 | - | 4 - 5 | 748 | 100 | |
BC118117.1 | - | 4 - 5 | 752 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |