Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TANGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035328.2 | NP_001030405.2 | 7 - 8 | 747 | 93 | |
GenBank mRNA | BC113297.1 | - | 7 - 8 | 747 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |