Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045886.2 | NP_001039351.1 | 8 - 9 | 785 | 68 | |
GenBank mRNA | BC105465.1 | - | 8 - 9 | 786 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |