Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGREF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045977.2 | NP_001039442.1 | 2 - 3 | 110 | 58 | |
GenBank mRNA | BC114163.1 | - | 2 - 3 | 111 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |