Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045998.2 | NP_001039463.1 | 5 - 6 | 577 | 111 | |
GenBank mRNA | BC105453.1 | - | 5 - 6 | 578 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |