Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046020.2 | NP_001039485.1 | 3 - 4 | 566 | 97 | |
GenBank mRNA | BC114047.1 | - | 3 - 4 | 569 | 97 | |
FJ594406.1 | - | 2 - 3 | 276 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |