Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046146.1 | NP_001039611.1 | 6 - 7 | 616 | 67 | |
GenBank mRNA | BC113208.1 | - | 6 - 7 | 616 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |