Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046148.1 | NP_001039613.1 | 10 - 11 | 1068 | 67 | |
GenBank mRNA | BC112777.1 | - | 10 - 11 | 1068 | 67 | |
BT029888.1 | - | 10 - 11 | 1065 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |