Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046162.1 | NP_001039627.1 | 6 - 7 | 1006 | 128 | |
GenBank mRNA | BC114034.1 | - | 6 - 7 | 1006 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |