Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1BG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046243.2 | NP_001039708.1 | 3 - 4 | 402 | 66 | |
GenBank mRNA | BC105374.1 | - | 3 - 4 | 405 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |