Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046427.2 | NP_001039892.1 | 6 - 7 | 417 | 122 | |
GenBank mRNA | BC112679.1 | - | 6 - 7 | 418 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |