Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046530.2 | NP_001039995.2 | 3 - 4 | 381 | 141 | |
GenBank mRNA | BC114134.1 | - | 3 - 4 | 382 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |