Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075142.1 | NP_001068610.1 | 6 - 7 | 723 | 90 | |
GenBank mRNA | AF081273.1 | - | 4 - 5 | 582 | 90 | |
BC118151.1 | - | 6 - 7 | 723 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |