Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H19orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075380.1 | NP_001068848.1 | 1 - 2 | 357 | 73 | |
GenBank mRNA | BC116031.1 | - | 1 - 2 | 357 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |