Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075426.1 | NP_001068894.1 | 2 - 3 | 1142 | 79 | |
GenBank mRNA | BC114763.1 | - | 2 - 3 | 1142 | 79 | |
EU599224.1 | - | 2 - 3 | 874 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |