Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075479.1 | NP_001068947.1 | 6 - 7 | 700 | 153 | |
GenBank mRNA | BC116035.1 | - | 6 - 7 | 700 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |