Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075552.1 | NP_001069020.1 | 5 - 6 | 686 | 59 | |
GenBank mRNA | BC114715.1 | - | 5 - 6 | 686 | 59 | |
BT025449.1 | - | 5 - 6 | 703 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |