Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075745.2 | NP_001069213.1 | 5 - 6 | 541 | 76 | |
GenBank mRNA | BC114010.1 | - | 5 - 6 | 542 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |