Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075803.2 | NP_001069271.1 | 9 - 10 | 1418 | 72 | |
GenBank mRNA | BC118450.1 | - | 9 - 10 | 1419 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |