Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075902.2 | NP_001069370.2 | 7 - 8 | 579 | 123 | |
GenBank mRNA | BC114844.1 | - | 7 - 8 | 580 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |