Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080217.1 | NP_001073686.1 | 3 - 4 | 263 | 70 | |
GenBank mRNA | AY173052.1 | - | 2 - 3 | 193 | 70 | |
BC113263.1 | - | 3 - 4 | 263 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |