Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAICS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205813.2 | NP_991382.2 | 7 - 8 | 1165 | 84 | |
GenBank mRNA | AY344127.1 | - | 8 - 9 | 1041 | 84 | |
BC113344.1 | - | 7 - 8 | 1165 | 84 | ||
BT030591.2 | - | 7 - 8 | 1007 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |