Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038198.2 | NP_001033287.1 | 5 - 6 | 1338 | 97 | |
GenBank mRNA | AY644769.1 | - | 3 - 4 | 1119 | 97 | |
AY644770.1 | - | 3 - 4 | 1122 | 97 | ||
BC123390.1 | - | 5 - 6 | 1338 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |