Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075134.1 | NP_001068602.1 | 6 - 7 | 816 | 95 | |
GenBank mRNA | BC119870.1 | - | 6 - 7 | 816 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |