Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBOAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075152.1 | NP_001068620.1 | 6 - 7 | 1103 | 119 | |
GenBank mRNA | BC119932.1 | - | 6 - 7 | 1103 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |