Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075236.1 | NP_001068704.1 | 19 - 20 | 1739 | 150 | |
GenBank mRNA | BC122692.1 | - | 19 - 20 | 1739 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |